Assay Details
Target Gene Details
Entrez Gene ID: | 90332 |
Gene Name: | exocyst complex component 3 like 2 |
Gene Aliases: |
XTP7 |
Location: |
Chr.19:45212621-45234211 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOC3L2 | NM_138568.3 | NP_612635.3 | ||
| AK093466.1 | BAC04175.1 | |||
| AK225791.1 | ||||
| BC104947.1 | AAI04948.1 | |||
| BC104949.1 | AAI04950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422482 | Chr.19:45173447 - 45266547 on Build GRCh38 | Duplication |
|
| esv3583434 | Chr.19:45185375 - 45248399 on Build GRCh38 | Loss |
|
| esv2678049 | Chr.19:45225268 - 45226944 on Build GRCh38 | Deletion |
|
| nsv833844 | Chr.19:45181051 - 45384148 on Build GRCh38 | Loss |
|
| nsv833843 | Chr.19:45090630 - 45290424 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map