Assay Details
Target Gene Details
Entrez Gene ID: | 6813 |
Gene Name: | syntaxin binding protein 2 |
Gene Aliases: |
FHL5, Hunc18b, MUNC18-2, UNC18-2, UNC18B, pp10122 |
Location: |
Chr.19:7637101-7647874 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STXBP2 | NM_001127396.2 | NP_001120868.1 | ||
| NM_001272034.1 | NP_001258963.1 | |||
| NM_006949.3 | NP_008880.2 | |||
| NR_073560.1 | ||||
| XM_011528210.1 | XP_011526512.1 | |||
| XM_011528212.2 | XP_011526514.1 | |||
| AB002559.1 | BAA19547.1 | |||
| AK222967.1 | BAD96687.1 | |||
| AK222977.1 | BAD96697.1 | |||
| AK302257.1 | ||||
| AK303701.1 | ||||
| AK314230.1 | ||||
| AU098728.1 | ||||
| BC002869.2 | AAH02869.1 | |||
| BQ067493.1 | ||||
| BT006915.1 | AAP35561.1 | |||
| DA431480.1 | ||||
| U63533.1 | AAC50762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953958 | Chr.19:7584715 - 7650814 on Build GRCh38 | Deletion |
|
| nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss |
|
| nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain |
|
| esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116037039,rs116588922] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map