Assay Details
Target Gene Details
Entrez Gene ID: | 6929 |
Gene Name: | transcription factor 3 |
Gene Aliases: |
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21 |
Location: |
Chr.19:1609290-1652590 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss |
|
| nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map