Assay Details
Target Gene Details
Entrez Gene ID: | 645432 |
Gene Name: | arrestin domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.19:4867614-4915469 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC5 | NM_001080523.1 | 2 | 370 | NP_001073992.1 |
| XM_005259616.2 | XP_005259673.1 | |||
| XM_011528181.2 | XP_011526483.1 | |||
| XM_017027144.1 | 2 | 584 | XP_016882633.1 | |
| XM_017027145.1 | 2 | 584 | XP_016882634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470117 | Chr.19:4811659 - 4900672 on Build GRCh38 | Loss |
|
| esv3893158 | Chr.19:4856682 - 4900672 on Build GRCh38 | Loss |
|
| nsv1065556 | Chr.19:4844193 - 4945238 on Build GRCh38 | Loss |
|
| esv25954 | Chr.19:4885251 - 4900646 on Build GRCh38 | Gain |
|

Genomic Map