Assay Details
Target Gene Details
Entrez Gene ID: | 147727 |
Gene Name: | ILF3 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.19:10651862-10653872 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILF3-AS1 | NR_024333.1 | 1 | 652 | |
| AF439744.2 | 1 | 262 | AAR01499.1 | |
| AK075016.1 | 1 | 619 | ||
| AK311360.1 | 1 | 621 | ||
| AL832185.1 | 1 | 625 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|

Genomic Map