Assay Details
Target Gene Details
Entrez Gene ID: | 125931 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 20 |
Gene Aliases: |
UNQ9366 |
Location: |
Chr.19:44503838-44530100 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM20 | XM_011526429.2 | 13 | 2745 | XP_011524731.1 |
| XM_011526430.2 | 12 | 2710 | XP_011524732.1 | |
| XM_011526432.2 | 9 | 1409 | XP_011524734.1 | |
| AY358129.1 | AAQ88496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061350 | Chr.19:44364917 - 44531444 on Build GRCh38 | Loss |
|

Genomic Map