Assay Details
Target Gene Details
Entrez Gene ID: | 634 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 1 |
Gene Aliases: |
BGP, BGP1, BGPI |
Location: |
Chr.19:42507306-42528509 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM1 | NM_001024912.2 | NP_001020083.1 | ||
| NM_001184813.1 | NP_001171742.1 | |||
| NM_001184815.1 | NP_001171744.1 | |||
| NM_001184816.1 | NP_001171745.1 | |||
| NM_001205344.1 | NP_001192273.1 | |||
| NM_001712.4 | NP_001703.2 | |||
| XM_011527206.1 | XP_011525508.1 | |||
| AK314211.1 | ||||
| AL833584.1 | ||||
| AY766113.1 | AAV34600.1 | |||
| BC014473.1 | AAH14473.1 | |||
| BC024164.1 | AAH24164.1 | |||
| BX485906.1 | ||||
| D12502.1 | BAA02063.1 | |||
| D90311.1 | BAA14341.1 | |||
| D90312.1 | BAA14342.1 | |||
| D90313.1 | BAA14343.1 | |||
| J03858.1 | AAA51826.1 | |||
| M69176.1 | AAA51825.1 | |||
| M72238.1 | AAA58393.1 | |||
| M76742.1 | AAA57142.1 | |||
| S71326.1 | AAB31183.1 | |||
| X14831.1 | CAA32940.1 | |||
| X16354.1 | CAA34404.1 | |||
| X16356.1 | CAA34405.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPE-AS1 | NR_073179.1 | |||
| NR_073180.1 | ||||
| AK096849.1 | BAC04875.1 | |||
| AK310497.1 | ||||
| AK311181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain |
|
| esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion |
|
| dgv15e196 | Chr.19:42487137 - 43165333 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map