Assay Details
Target Gene Details
Entrez Gene ID: | 23031 |
Gene Name: | microtubule associated serine/threonine kinase 3 |
Gene Aliases: |
- |
Location: |
Chr.19:18097764-18151692 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAST3 | NM_015016.1 | NP_055831.1 | ||
| XM_005259824.2 | XP_005259881.1 | |||
| XM_005259825.2 | XP_005259882.1 | |||
| XM_006722698.2 | XP_006722761.1 | |||
| XM_006722699.2 | XP_006722762.2 | |||
| XM_011527823.1 | XP_011526125.1 | |||
| XM_017026506.1 | XP_016881995.1 | |||
| XM_017026507.1 | XP_016881996.1 | |||
| XM_017026508.1 | XP_016881997.1 | |||
| XM_017026509.1 | XP_016881998.1 | |||
| XM_017026510.1 | XP_016881999.1 | |||
| XM_017026511.1 | XP_016882000.1 | |||
| XM_017026512.1 | XP_016882001.1 | |||
| XM_017026513.1 | XP_016882002.1 | |||
| XM_017026514.1 | XP_016882003.1 | |||
| XM_017026515.1 | XP_016882004.1 | |||
| XM_017026516.1 | XP_016882005.1 | |||
| AB011133.1 | BAA25487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160592 | Chr.19:18100050 - 18194768 on Build GRCh38 | Deletion |
|
| nsv833775 | Chr.19:18102951 - 18279264 on Build GRCh38 | Loss |
|
| nsv470130 | Chr.19:17936474 - 18123777 on Build GRCh38 | Loss |
|
| nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map