Assay Details
Target Gene Details
Entrez Gene ID: | 10498 |
Gene Name: | coactivator associated arginine methyltransferase 1 |
Gene Aliases: |
PRMT4 |
Location: |
Chr.19:10871577-10923078 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARM1 | NM_199141.1 | NP_954592.1 | ||
| XM_005259708.4 | XP_005259765.1 | |||
| AL833242.1 | ||||
| BU176535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss |
|
| nsv833748 | Chr.19:10757002 - 10925368 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map