Assay Details
Target Gene Details
Entrez Gene ID: | 5585 |
Gene Name: | protein kinase N1 |
Gene Aliases: |
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1 |
Location: |
Chr.19:14433354-14471867 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKN1 | NM_002741.3 | 6 | 1107 | NP_002732.3 |
| NM_213560.1 | 6 | 974 | NP_998725.1 | |
| XM_011528128.1 | 6 | 990 | XP_011526430.1 | |
| AK123007.1 | 6 | 988 | ||
| AK292130.1 | 6 | 971 | ||
| AK313886.1 | 6 | 947 | ||
| BC040061.1 | 6 | 1104 | AAH40061.1 | |
| BC094766.1 | 6 | 1138 | AAH94766.1 | |
| D26181.1 | 6 | 975 | BAA05169.1 | |
| S75546.1 | 6 | 1023 | AAB33345.1 | |
| U33053.1 | 6 | 1023 | AAC50209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470127 | Chr.19:14434730 - 14479800 on Build GRCh38 | Loss |
|
| nsv1160590 | Chr.19:14398750 - 14462842 on Build GRCh38 | Deletion |
|
| nsv833764 | Chr.19:14381787 - 14504061 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map