Assay Details
Target Gene Details
Entrez Gene ID: | 388551 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 16 |
Gene Aliases: |
CEAL2, DFNA4B |
Location: |
Chr.19:44699151-44710714 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM16 | NM_001039213.3 | NP_001034302.2 | ||
| XM_017026795.1 | 1 | 626 | XP_016882284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9737 | Chr.19:44674838 - 44703359 on Build GRCh38 | Gain |
|
| nsv1062812 | Chr.19:44538275 - 44711525 on Build GRCh38 | Gain |
|
| nsv483005 | Chr.19:44530751 - 44705087 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map