Assay Details
Target Gene Details
Entrez Gene ID: | 2139 |
Gene Name: | EYA transcriptional coactivator and phosphatase 2 |
Gene Aliases: |
EAB1 |
Location: |
Chr.20:46894624-47188844 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA2 | NM_005244.4 | NP_005235.3 | ||
| NM_172110.3 | NP_742108.2 | |||
| XM_005260327.2 | XP_005260384.1 | |||
| XM_017027721.1 | XP_016883210.1 | |||
| AF055015.1 | AAC09362.1 | |||
| AK093582.1 | ||||
| AK293032.1 | ||||
| AY705349.1 | AAU11525.1 | |||
| BC000289.2 | AAH00289.2 | |||
| BC008803.2 | AAH08803.1 | |||
| BC013882.2 | AAH13882.2 | |||
| BT006682.1 | AAP35328.1 | |||
| U71207.1 | AAB51120.1 | |||
| U81601.1 | AAB42065.1 |
Target Gene Details
Entrez Gene ID: | 100500814 |
Gene Name: | microRNA 3616 |
Gene Aliases: |
mir-3616 |
Location: |
Chr.20:47166967-47167058 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR3616 | NR_037410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33497 | Chr.20:47143439 - 47650962 on Build GRCh38 | Loss |
|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78308190] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map