Assay Details
Target Gene Details
Entrez Gene ID: | 101927117 |
Gene Name: | PLCG1 antisense RNA 1 |
Gene Aliases: |
TOP1-AS1 |
Location: |
Chr.20:41097994-41138000 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCG1-AS1 | NR_109889.1 |
Target Gene Details
Entrez Gene ID: | 7150 |
Gene Name: | topoisomerase (DNA) I |
Gene Aliases: |
TOPI |
Location: |
Chr.20:41028822-41124487 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOP1 | NM_003286.2 | NP_003277.1 | ||
| AK292943.1 | ||||
| BC000943.2 | AAH00943.1 | |||
| BC136297.1 | ||||
| J03250.1 | AAA61207.1 | |||
| U07804.1 | AAB60379.1 | |||
| U07806.1 | AAB60380.1 | |||
| X16479.2 | CAA34500.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064246 | Chr.20:40261343 - 42254320 on Build GRCh38 | Loss |
|
| nsv833983 | Chr.20:41051961 - 41213015 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map