Assay Details
Target Gene Details
Entrez Gene ID: | 140862 |
Gene Name: | isthmin 1, angiogenesis inhibitor |
Gene Aliases: |
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1 |
Location: |
Chr.20:13221771-13326734 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISM1 | NM_080826.1 | NP_543016.1 | ||
| XM_017027680.1 | XP_016883169.1 | |||
| DB169194.1 |
Target Gene Details
Entrez Gene ID: | 55617 |
Gene Name: | taspase 1 |
Gene Aliases: |
C20orf13, dJ585I14.2 |
Location: |
Chr.20:13104760-13638936 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TASP1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4247n100 | Chr.20:12748832 - 13357690 on Build GRCh38 | Gain |
|
| nsv518184 | Chr.20:12763161 - 13364025 on Build GRCh38 | Gain |
|
| esv3584449 | Chr.20:13218706 - 13235785 on Build GRCh38 | Loss |
|
| nsv952987 | Chr.20:13219254 - 13222853 on Build GRCh38 | Deletion |
|

Genomic Map