Assay Details
Target Gene Details
Entrez Gene ID: | 8888 |
Gene Name: | minichromosome maintenance complex component 3 associated protein |
Gene Aliases: |
GANP, MAP80, SAC3 |
Location: |
Chr.21:46235125-46286274 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM3AP | XM_005261203.4 | 1 | 65 | XP_005261260.1 |
| XM_005261204.4 | 1 | 65 | XP_005261261.1 | |
| XM_005261205.3 | XP_005261262.1 | |||
| HF584748.1 |
Target Gene Details
Entrez Gene ID: | 54059 |
Gene Name: | ybeY metallopeptidase (putative) |
Gene Aliases: |
C21orf57 |
Location: |
Chr.21:46286056-46314188 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YBEY | XM_005261156.3 | 1 | 155 | XP_005261213.2 |
| XM_011529634.2 | 1 | 155 | XP_011527936.2 | |
| XM_017028394.1 | 1 | 155 | XP_016883883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv3547 | Chr.21:46283787 - 46321516 on Build GRCh38 | Insertion |
|
| nsv527355 | Chr.21:46258860 - 46443112 on Build GRCh38 | Gain |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv587975 | Chr.21:46241321 - 46410929 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|
| nsv587977 | Chr.21:46284982 - 46293172 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map