Assay Details
Target Gene Details
Entrez Gene ID: | 84536 |
Gene Name: | long intergenic non-protein coding RNA 1547 |
Gene Aliases: |
C21orf67, C21orf69, PRED54 |
Location: |
Chr.21:44933284-44939913 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01547 | NR_027128.1 | |||
| NR_027129.1 | ||||
| AF380178.1 | AAL34496.1 | |||
| AK127059.1 | 1 | 1005 | ||
| AY040088.1 | AAK74136.1 | |||
| BI829356.1 | ||||
| BI907880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647138 | Chr.21:44916487 - 44942284 on Build GRCh38 | Loss |
|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|

Genomic Map