Assay Details
Target Gene Details
Entrez Gene ID: | 3275 |
Gene Name: | protein arginine methyltransferase 2 |
Gene Aliases: |
HRMT1L1 |
Location: |
Chr.21:46635153-46665685 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587999 | Chr.21:46522003 - 46683939 on Build GRCh38 | Gain |
|
| dgv4442n100 | Chr.21:46551020 - 46677033 on Build GRCh38 | Gain |
|
| dgv4444n100 | Chr.21:46599898 - 46677033 on Build GRCh38 | Gain |
|
| nsv1059706 | Chr.21:46627762 - 46662768 on Build GRCh38 | Gain |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| esv3647171 | Chr.21:46586178 - 46674643 on Build GRCh38 | Gain |
|
| dgv4441n100 | Chr.21:46474792 - 46677033 on Build GRCh38 | Gain |
|
| dgv4449n100 | Chr.21:46634852 - 46677033 on Build GRCh38 | Gain+Loss |
|
| dgv4448n100 | Chr.21:46620242 - 46677033 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map