Assay Details
Target Gene Details
Entrez Gene ID: | 150084 |
Gene Name: | immunoglobulin superfamily member 5 |
Gene Aliases: |
GSF5, JAM4 |
Location: |
Chr.21:39725838-39802096 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF5 | XM_011529472.1 | 2 | 1111 | XP_011527774.1 |
| XM_011529473.1 | 2 | 1111 | XP_011527775.1 | |
| XM_017028279.1 | 2 | 1111 | XP_016883768.1 | |
| XM_017028280.1 | 2 | 1111 | XP_016883769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834099 | Chr.21:39704946 - 39834103 on Build GRCh38 | Loss |
|
| nsv1061454 | Chr.21:39386047 - 39893479 on Build GRCh38 | Gain |
|
| esv3647026 | Chr.21:39723451 - 39731234 on Build GRCh38 | Loss |
|
| esv2660100 | Chr.21:39723462 - 39731242 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map