Assay Details
Target Gene Details
Entrez Gene ID: | 23181 |
Gene Name: | disco interacting protein 2 homolog A |
Gene Aliases: |
C21orf106, DIP2 |
Location: |
Chr.21:46458942-46570900 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 100862692 |
Gene Name: | DIP2A intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.21:46462471-46469306 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIP2A-IT1 | NR_046400.1 | 4 | 320 | |
| BM929894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762115 | Chr.21:46465226 - 46475517 on Build GRCh38 | Loss |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv587998 | Chr.21:46436193 - 46565742 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map