Assay Details
Target Gene Details
Entrez Gene ID: | 85395 |
Gene Name: | family with sequence similarity 207 member A |
Gene Aliases: |
C21orf70, PRED56 |
Location: |
Chr.21:44939697-44976989 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM207A | NM_001316983.1 | NP_001303912.1 | ||
| NM_001316984.1 | NP_001303913.1 | |||
| NM_001316985.1 | NP_001303914.1 | |||
| NM_001316986.1 | NP_001303915.1 | |||
| NM_001316987.1 | NP_001303916.1 | |||
| NM_001316988.1 | NP_001303917.1 | |||
| NM_058190.3 | NP_478070.1 | |||
| XM_017028481.1 | XP_016883970.1 | |||
| AF391113.1 | AAL34504.1 | |||
| AF391114.1 | AAL34505.1 | |||
| BC009341.2 | AAH09341.1 | |||
| BG479572.1 | ||||
| BG744869.1 | ||||
| BM042090.1 | ||||
| CN368049.1 | ||||
| CR998572.1 | ||||
| HY097926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|
| nsv953649 | Chr.21:44943186 - 44960585 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs572333263] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map