Assay Details
Target Gene Details
Entrez Gene ID: | 3275 |
Gene Name: | protein arginine methyltransferase 2 |
Gene Aliases: |
HRMT1L1 |
Location: |
Chr.21:46635153-46665685 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT2 | NM_001242864.2 | NP_001229793.1 | ||
| NM_001242866.2 | 7 | 2472 | NP_001229795.1 | |
| NM_001286676.1 | NP_001273605.1 | |||
| NM_001286677.1 | NP_001273606.1 | |||
| NM_001286678.1 | NP_001273607.1 | |||
| NM_001535.4 | NP_001526.2 | |||
| NM_206962.3 | NP_996845.1 | |||
| XM_005261111.3 | XP_005261168.1 | |||
| XM_006723998.3 | XP_006724061.1 | |||
| XM_006724000.2 | XP_006724063.1 | |||
| AK095641.1 | ||||
| AK123352.1 | ||||
| AK123650.1 | ||||
| AK124494.1 | ||||
| AK223157.1 | BAD96877.1 | |||
| AK312573.1 | ||||
| AL833586.1 | ||||
| BC000727.2 | AAH00727.1 | |||
| BC038765.1 | AAH38765.1 | |||
| BC100026.1 | AAI00027.1 | |||
| BY799824.2 | 1 | 263 | ||
| CR541804.1 | CAG46603.1 | |||
| FJ436410.1 | ||||
| FJ436411.1 | ||||
| FJ436412.1 | ||||
| KU178127.1 | ||||
| KU178128.1 | ||||
| KU178129.1 | ||||
| U80213.1 | AAB48437.1 | |||
| X99209.1 | CAA67599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587999 | Chr.21:46522003 - 46683939 on Build GRCh38 | Gain |
|
| dgv4442n100 | Chr.21:46551020 - 46677033 on Build GRCh38 | Gain |
|
| dgv4444n100 | Chr.21:46599898 - 46677033 on Build GRCh38 | Gain |
|
| nsv1059706 | Chr.21:46627762 - 46662768 on Build GRCh38 | Gain |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| esv3647171 | Chr.21:46586178 - 46674643 on Build GRCh38 | Gain |
|
| dgv4441n100 | Chr.21:46474792 - 46677033 on Build GRCh38 | Gain |
|
| dgv4448n100 | Chr.21:46620242 - 46677033 on Build GRCh38 | Gain |
|
| dgv4449n100 | Chr.21:46634852 - 46677033 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map