Assay Details
Target Gene Details
Entrez Gene ID: | 54059 |
Gene Name: | ybeY metallopeptidase (putative) |
Gene Aliases: |
C21orf57 |
Location: |
Chr.21:46286056-46314188 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YBEY | NM_001314022.1 | NP_001300951.1 | ||
| NM_001314023.1 | NP_001300952.1 | |||
| NM_001314024.1 | NP_001300953.1 | |||
| NM_001314025.1 | NP_001300954.1 | |||
| NM_058181.2 | NP_478061.1 | |||
| XM_005261156.3 | XP_005261213.2 | |||
| XM_011529629.2 | XP_011527931.1 | |||
| XM_011529630.2 | XP_011527932.1 | |||
| XM_011529631.2 | XP_011527933.1 | |||
| XM_011529633.2 | XP_011527935.1 | |||
| XM_011529634.2 | XP_011527936.2 | |||
| XM_011529635.2 | XP_011527937.1 | |||
| XM_017028394.1 | XP_016883883.1 | |||
| AI262212.1 | ||||
| AK128875.1 | ||||
| AY040873.1 | AAK74138.1 | |||
| AY040874.1 | AAK74139.1 | |||
| AY040875.1 | AAK74140.1 | |||
| AY040876.1 | AAK74141.1 | |||
| BG473705.1 | ||||
| BM796288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv3547 | Chr.21:46283787 - 46321516 on Build GRCh38 | Insertion |
|
| nsv527355 | Chr.21:46258860 - 46443112 on Build GRCh38 | Gain |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv587975 | Chr.21:46241321 - 46410929 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map