Assay Details
Target Gene Details
Entrez Gene ID: | 3689 |
Gene Name: | integrin subunit beta 2 |
Gene Aliases: |
CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7 |
Location: |
Chr.21:44885949-44928873 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB2 | NM_001127491.2 | NP_001120963.2 | ||
| AK095992.1 | ||||
| AU279404.1 | ||||
| DA200537.1 |
Target Gene Details
Entrez Gene ID: | 100505746 |
Gene Name: | ITGB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.21:44921035-44929680 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB2-AS1 | NR_038311.1 | |||
| NR_038312.1 | ||||
| NR_038313.1 | ||||
| NR_038314.1 | ||||
| NR_038315.1 | ||||
| NR_038316.1 | ||||
| DA378472.1 | ||||
| DA942354.1 | ||||
| DA944688.1 | ||||
| DA948979.1 | ||||
| DB115909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575398 | Chr.21:44921339 - 44924818 on Build GRCh38 | Gain |
|
| esv3647138 | Chr.21:44916487 - 44942284 on Build GRCh38 | Loss |
|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map