Assay Details
Target Gene Details
Entrez Gene ID: | 64699 |
Gene Name: | transmembrane protease, serine 3 |
Gene Aliases: |
DFNB10, DFNB8, ECHOS1, TADG12 |
Location: |
Chr.21:42371887-42396846 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS3 | NM_001256317.1 | NP_001243246.1 | ||
| NM_024022.2 | NP_076927.1 | |||
| NM_032404.2 | NP_115780.1 | |||
| NR_046020.1 | ||||
| AB038157.1 | BAB20077.1 | |||
| AB038158.1 | BAB20078.1 | |||
| AB038159.1 | BAB20079.1 | |||
| AF201380.1 | AAG37012.1 | |||
| AK131212.1 | ||||
| AK172842.1 | BAD18806.1 | |||
| AY358458.1 | AAQ88823.1 | |||
| AY633572.1 | AAT66641.1 | |||
| BC074846.2 | AAH74846.1 | |||
| BC074847.2 | AAH74847.1 |
More Information
Set Membership: |
|

Genomic Map