Assay Details
Target Gene Details
Entrez Gene ID: | 85395 |
Gene Name: | family with sequence similarity 207 member A |
Gene Aliases: |
C21orf70, PRED56 |
Location: |
Chr.21:44939697-44976989 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM207A | NM_001316988.1 | NP_001303917.1 | ||
| BG744869.1 |
Target Gene Details
Entrez Gene ID: | 84536 |
Gene Name: | long intergenic non-protein coding RNA 1547 |
Gene Aliases: |
C21orf67, C21orf69, PRED54 |
Location: |
Chr.21:44933284-44939913 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01547 | NR_027128.1 | 1 | 59 | |
| NR_027129.1 | 1 | 59 | ||
| AF380178.1 | AAL34496.1 | |||
| AK127059.1 | 1 | 59 | ||
| AY040088.1 | AAK74136.1 | |||
| BI829356.1 | ||||
| BI907880.1 | 1 | 59 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647138 | Chr.21:44916487 - 44942284 on Build GRCh38 | Loss |
|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76091017] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map