Assay Details
Target Gene Details
Entrez Gene ID: | 5905 |
Gene Name: | Ran GTPase activating protein 1 |
Gene Aliases: |
Fug1, RANGAP, SD |
Location: |
Chr.22:41244777-41302213 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459888 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain |
|
| nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss |
|
| nsv817997 | Chr.22:41244293 - 41366866 on Build GRCh38 | Gain |
|
| nsv589049 | Chr.22:41168704 - 41276267 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144019704] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map