Assay Details
Target Gene Details
Entrez Gene ID: | 129138 |
Gene Name: | ankyrin repeat domain 54 |
Gene Aliases: |
LIAR |
Location: |
Chr.22:37830854-37848994 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD54 | XM_011529878.2 | 1 | 725 | XP_011528180.1 |
| XM_017028590.1 | 1 | 725 | XP_016884079.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
| esv3893482 | Chr.22:37832559 - 37927500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map