Assay Details
Target Gene Details
Entrez Gene ID: | 553158 |
Gene Name: | PRR5-ARHGAP8 readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:44702198-44862784 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR5-ARHGAP8 | NM_181334.5 | NP_851851.3 | ||
| AF195968.1 | AAK58136.1 | |||
| AF195969.1 | AAK58137.1 | |||
| AL355192.1 | CAB90248.1 | |||
| BC132755.1 | ||||
| BC132757.1 | ||||
| BC144382.1 | ||||
| BX335168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1325e212 | Chr.22:44734370 - 44741324 on Build GRCh38 | Loss |
|
| nsv589108 | Chr.22:44721714 - 44796741 on Build GRCh38 | Loss |
|
| nsv9918 | Chr.22:44738952 - 44743076 on Build GRCh38 | Loss |
|
| nsv589109 | Chr.22:44725984 - 44739384 on Build GRCh38 | Loss |
|
| nsv829268 | Chr.22:44640618 - 44785937 on Build GRCh38 | Gain |
|
| esv2758842 | Chr.22:44630924 - 44814649 on Build GRCh38 | Loss |
|
| dgv4586n100 | Chr.22:44719617 - 44799761 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114560248,rs75368138] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map