Assay Details
Target Gene Details
Entrez Gene ID: | 100130418 |
Gene Name: | cat eye syndrome chromosome region, candidate 7 (non-protein coding) |
Gene Aliases: |
SAHL1 |
Location: |
Chr.22:17036570-17058792 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR7 | NR_015352.1 | 4 | 2289 | |
| AK098045.1 | 4 | 1620 | ||
| AY026052.1 | 4 | 2289 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4460n100 | Chr.22:16384224 - 17135620 on Build GRCh38 | Gain |
|
| esv3647204 | Chr.22:16939332 - 17123705 on Build GRCh38 | Gain |
|
| dgv4463n100 | Chr.22:16749141 - 17081023 on Build GRCh38 | Gain |
|
| nsv1067241 | Chr.22:16757380 - 17351252 on Build GRCh38 | Loss |
|

Genomic Map