Assay Details
Target Gene Details
Entrez Gene ID: | 102724378 |
Gene Name: | uncharacterized LOC102724378 |
Gene Aliases: |
- |
Location: |
Chr.22:37738726-37747457 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724378 | XM_011530647.2 | XP_011528949.1 | ||
| XM_017029173.1 | XP_016884662.1 |
Target Gene Details
Entrez Gene ID: | 11078 |
Gene Name: | TRIO and F-actin binding protein |
Gene Aliases: |
DFNB28, HRIHFB2122, TAP68, TARA, dJ37E16.4 |
Location: |
Chr.22:37696988-37776556 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIOBP | NM_001039141.2 | NP_001034230.1 | ||
| AK096634.2 | ||||
| DQ228003.1 | ABB59559.1 | |||
| DQ228005.1 | ABB59561.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829203 | Chr.22:36998126 - 37773322 on Build GRCh38 | Loss |
|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
| esv3575478 | Chr.22:37722215 - 37791928 on Build GRCh38 | Gain |
|
| nsv471197 | Chr.22:37600773 - 37762734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map