Assay Details
Target Gene Details
Entrez Gene ID: | 57553 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 3 |
Gene Aliases: |
MICAL-3 |
Location: |
Chr.22:17787649-18024559 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509806 | Chr.22:17806210 - 17883749 on Build GRCh38 | Insertion |
|
| nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain |
|
| esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss |
|
| nsv834120 | Chr.22:17733890 - 17904438 on Build GRCh38 | Loss |
|
| nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map