Assay Details
Target Gene Details
Entrez Gene ID: | 23331 |
Gene Name: | tetratricopeptide repeat domain 28 |
Gene Aliases: |
TPRBK |
Location: |
Chr.22:27978014-28679865 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC28 | NM_001145418.1 | 23 | 7839 | NP_001138890.1 |
| XM_005261405.2 | 22 | 7815 | XP_005261462.1 | |
| XM_006724171.3 | 22 | 16617 | XP_006724234.1 | |
| XM_011530018.2 | 23 | 8018 | XP_011528320.1 | |
| XM_017028673.1 | 23 | 7812 | XP_016884162.1 | |
| AB028966.3 | 18 | 6632 | BAA82995.3 | |
| AB665284.1 | 23 | 7787 | ||
| AL080174.1 | 2 | 1970 | CAB45760.2 | |
| AL137328.1 | 1 | 607 | CAB70697.1 | |
| BC016465.1 | 15 | 4383 | AAH16465.1 |
Target Gene Details
Entrez Gene ID: | 284900 |
Gene Name: | TTC28 antisense RNA 1 |
Gene Aliases: |
TTC28-AS, TTC28AS, dJ353E16.2 |
Location: |
Chr.22:27919376-28002679 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC28-AS1 | NR_026963.1 | |||
| AL832597.1 | ||||
| DA934139.1 | ||||
| DB254752.1 |
More Information
Set Membership: |
|

Genomic Map