Assay Details
Target Gene Details
Entrez Gene ID: | 55615 |
Gene Name: | proline rich 5 |
Gene Aliases: |
FLJ20185k, PP610, PROTOR-1, PROTOR1 |
Location: |
Chr.22:44668547-44737681 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR5 | NM_001017528.2 | NP_001017528.1 | ||
| NM_001017529.2 | NP_001017529.1 | |||
| NM_001017530.1 | NP_001017530.1 | |||
| NM_001198721.1 | NP_001185650.1 | |||
| NM_015366.3 | NP_056181.2 | |||
| AK055848.1 | ||||
| AK091884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3650 | Chr.22:44650253 - 44683874 on Build GRCh38 | Insertion |
|
| nsv829268 | Chr.22:44640618 - 44785937 on Build GRCh38 | Gain |
|
| nsv1160777 | Chr.22:44606668 - 44692090 on Build GRCh38 | Duplication |
|
| esv2758842 | Chr.22:44630924 - 44814649 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74641124] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map