Assay Details
Target Gene Details
Entrez Gene ID: | 9463 |
Gene Name: | protein interacting with PRKCA 1 |
Gene Aliases: |
PICK, PRKCABP |
Location: |
Chr.22:38056311-38075704 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PICK1 | XM_017029091.1 | 1 | 230 | XP_016884580.1 |
| XM_017029094.1 | 1 | 207 | XP_016884583.1 | |
| XM_017029095.1 | 1 | 207 | XP_016884584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss |
|
| nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map