Assay Details
Target Gene Details
Entrez Gene ID: | 9463 |
Gene Name: | protein interacting with PRKCA 1 |
Gene Aliases: |
PICK, PRKCABP |
Location: |
Chr.22:38056311-38075704 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PICK1 | NM_001039583.1 | NP_001034672.1 | ||
| NM_001039584.1 | NP_001034673.1 | |||
| NM_012407.3 | NP_036539.1 | |||
| XM_006724379.2 | XP_006724442.1 | |||
| XM_006724380.2 | XP_006724443.1 | |||
| XM_011530517.2 | XP_011528819.1 | |||
| XM_017029090.1 | XP_016884579.1 | |||
| XM_017029091.1 | XP_016884580.1 | |||
| XM_017029092.1 | XP_016884581.1 | |||
| XM_017029093.1 | XP_016884582.1 | |||
| XM_017029094.1 | XP_016884583.1 | |||
| XM_017029095.1 | XP_016884584.1 | |||
| AB026491.1 | BAA89294.1 | |||
| AF231710.1 | AAF97502.1 | |||
| AK000875.1 | ||||
| AK092818.1 | ||||
| AK222640.1 | BAD96360.1 | |||
| AK299684.1 | ||||
| AK308792.1 | ||||
| AK310629.1 | ||||
| AL049654.1 | CAB41082.1 | |||
| BC017561.2 | AAH17561.1 | |||
| CB141073.1 | ||||
| CN482899.1 | ||||
| CR456550.1 | CAG30436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471198 | Chr.22:38019714 - 38149612 on Build GRCh38 | Loss |
|
| nsv834193 | Chr.22:37997329 - 38172090 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map