Assay Details
Target Gene Details
Entrez Gene ID: | 1413 |
Gene Name: | crystallin beta A4 |
Gene Aliases: |
CTRCT23, CYRBA4, MCOPCT4 |
Location: |
Chr.22:26607982-26630672 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYBA4 | NM_001886.2 | NP_001877.1 | ||
| XM_006724140.3 | XP_006724203.1 | |||
| XM_017028598.1 | XP_016884087.1 | |||
| BC069404.1 | AAH69404.1 | |||
| BC096171.3 | AAH96171.1 | |||
| BC096172.3 | AAH96172.1 | |||
| BC096173.3 | AAH96173.1 | |||
| BC096174.1 | AAH96174.1 | |||
| BM667295.1 | ||||
| CR456424.1 | CAG30310.1 | |||
| KM230606.1 | ||||
| KM365088.1 | ||||
| KM365089.1 | ||||
| KM365090.1 | ||||
| U59057.1 | AAC50970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758831 | Chr.22:26502533 - 26633932 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map