Assay Details
Target Gene Details
Entrez Gene ID: | 2182 |
Gene Name: | acyl-CoA synthetase long-chain family member 4 |
Gene Aliases: |
ACS4, FACL4, LACS4, MRX63, MRX68 |
Location: |
Chr.X:109641330-109733403 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSL4 | NM_001318509.1 | NP_001305438.1 | ||
| NM_001318510.1 | NP_001305439.1 | |||
| NM_004458.2 | NP_004449.1 | |||
| NM_022977.2 | NP_075266.1 | |||
| XM_005262109.2 | XP_005262166.1 | |||
| XM_006724635.2 | XP_006724698.1 | |||
| XM_011530888.2 | XP_011529190.1 | |||
| XM_011530889.2 | XP_011529191.1 | |||
| AB061713.1 | BAB86900.1 | |||
| AB061714.1 | BAB86901.1 | |||
| AF030555.1 | AAC17493.1 | |||
| AK292070.1 | ||||
| AK294915.1 | ||||
| AK307566.1 | ||||
| BC034959.2 | AAH34959.1 | |||
| Y12777.1 | CAA73314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438149 | Chr.X:109652018 - 109730612 on Build GRCh38 | Loss |
|
| nsv523984 | Chr.X:109674544 - 109723196 on Build GRCh38 | Gain |
|
| nsv7034 | Chr.X:109673490 - 109713065 on Build GRCh38 | Deletion |
|
| esv2758883 | Chr.X:109535772 - 109737663 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map