Assay Details
Target Gene Details
Entrez Gene ID: | 331 |
Gene Name: | X-linked inhibitor of apoptosis |
Gene Aliases: |
API3, BIRC4, IAP-3, ILP1, MIHA, XLP2, hIAP-3, hIAP3 |
Location: |
Chr.X:123859812-123913979 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XIAP | NM_001167.3 | NP_001158.2 | ||
| NM_001204401.1 | NP_001191330.1 | |||
| NR_037916.1 | ||||
| XM_006724754.2 | XP_006724817.1 | |||
| XM_011531329.2 | XP_011529631.1 | |||
| AK309704.1 | ||||
| AK313883.1 | ||||
| BC032729.1 | AAH32729.1 | |||
| DA316362.1 | ||||
| U32974.1 | AAC50518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758890 | Chr.X:123830966 - 124028715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map