Assay Details
Target Gene Details
Entrez Gene ID: | 10479 |
Gene Name: | solute carrier family 9 member A6 |
Gene Aliases: |
MRSA, NHE6 |
Location: |
Chr.X:135974597-136047269 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A6 | XM_017029223.1 | XP_016884712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758895 | Chr.X:135468612 - 136072582 on Build GRCh38 | Gain+Loss |
|
| nsv1076276 | Chr.X:135973540 - 135976741 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map