Assay Details
Target Gene Details
Entrez Gene ID: | 91851 |
Gene Name: | chordin like 1 |
Gene Aliases: |
CHL, MGC1, MGCN, NRLN1, VOPT, dA141H5.1 |
Location: |
Chr.X:110673856-110796058 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRDL1 | NM_001143981.1 | NP_001137453.1 | ||
| NM_001143982.1 | NP_001137454.1 | |||
| NM_001143983.2 | NP_001137455.2 | |||
| NM_145234.3 | NP_660277.2 | |||
| XM_005262221.1 | XP_005262278.1 | |||
| XM_005262222.3 | XP_005262279.1 | |||
| XM_005262223.1 | XP_005262280.1 | |||
| XM_005262224.1 | XP_005262281.1 | |||
| XM_017029959.1 | XP_016885448.1 | |||
| AB209548.1 | BAD92785.1 | |||
| AK092245.1 | ||||
| AK293106.1 | ||||
| AK297563.1 | ||||
| AK312270.1 | ||||
| AY608914.1 | AAU25841.1 | |||
| BC002909.2 | AAH02909.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7037 | Chr.X:110684665 - 110729447 on Build GRCh38 | Deletion |
|
| nsv438151 | Chr.X:110695977 - 110697931 on Build GRCh38 | Loss |
|
| esv2666835 | Chr.X:110695373 - 110698911 on Build GRCh38 | Deletion |
|
| nsv515161 | Chr.X:110695500 - 110698324 on Build GRCh38 | Loss |
|
| esv21564 | Chr.X:110695400 - 110698973 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map