Assay Details
Target Gene Details
Entrez Gene ID: | 6839 |
Gene Name: | suppressor of variegation 3-9 homolog 1 |
Gene Aliases: |
H3-K9-HMTase 1, KMT1A, MG44, SUV39H |
Location: |
Chr.X:48695554-48709016 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUV39H1 | NM_001282166.1 | 1 | 55 | NP_001269095.1 |
| AK299900.1 | 1 | 55 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map