Assay Details
Target Gene Details
Entrez Gene ID: | 105373311 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.X:109732278-109734332 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373311 | XM_011531090.2 | 1 | 1616 | XP_011529392.1 |
| AK294197.1 | 1 | 1194 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758883 | Chr.X:109535772 - 109737663 on Build GRCh38 | Loss |
|

Genomic Map