Assay Details
Target Gene Details
Entrez Gene ID: | 6247 |
Gene Name: | retinoschisin 1 |
Gene Aliases: |
RS, XLRS1 |
Location: |
Chr.X:18639688-18672103 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RS1 | NM_000330.3 | NP_000321.1 | ||
| AF014459.1 | AAC17928.1 | |||
| BQ636812.1 | ||||
| DQ426892.1 |
More Information
Set Membership: |
|

Genomic Map