Assay Details
Target Gene Details
Entrez Gene ID: | 10159 |
Gene Name: | ATPase H+ transporting accessory protein 2 |
Gene Aliases: |
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS |
Location: |
Chr.X:40580964-40606637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6AP2 | NM_005765.2 | NP_005756.2 | ||
| AF109363.1 | AAQ13511.1 | |||
| AF248966.1 | AAG44564.1 | |||
| AF291814.1 | AAM47531.1 | |||
| AK075382.1 | BAC11582.1 | |||
| AK293510.1 | ||||
| AK296614.1 | ||||
| AK297007.1 | ||||
| AK297760.1 | ||||
| AK314633.1 | ||||
| AK315948.1 | ||||
| AY038990.1 | AAK83467.1 | |||
| BC084541.1 | AAH84541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470350 | Chr.X:40540044 - 40624703 on Build GRCh38 | Gain |
|
| nsv518965 | Chr.X:40569250 - 40754042 on Build GRCh38 | Gain |
|
| esv33829 | Chr.X:40117374 - 40692316 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map