Assay Details
Target Gene Details
Entrez Gene ID: | 100132618 |
Gene Name: | ZRANB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:71046506-71067184 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZRANB2-AS1 | NR_038420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546472 | Chr.1:70967301 - 71075320 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
