Assay Details
Target Gene Details
Entrez Gene ID: | 1379 |
Gene Name: | complement component 3b/4b receptor 1-like |
Gene Aliases: |
- |
Location: |
Chr.1:207645113-207723691 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CR1L | NM_175710.1 | NP_783641.1 | ||
| BC109190.1 | AAI09191.1 | |||
| BC109191.1 | AAI09192.1 | |||
| BX094042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891082 | Chr.1:207523372 - 207779899 on Build GRCh38 | Loss |
|
| esv3588675 | Chr.1:207667715 - 207672472 on Build GRCh38 | Loss |
|
| esv25546 | Chr.1:207667857 - 207672436 on Build GRCh38 | Loss |
|
| esv2675458 | Chr.1:207540634 - 207708875 on Build GRCh38 | Deletion |
|
| esv2722106 | Chr.1:207502410 - 207681369 on Build GRCh38 | Deletion |
|
| esv3588674 | Chr.1:207630455 - 207727138 on Build GRCh38 | Gain |
|
| esv2676306 | Chr.1:207667717 - 207672469 on Build GRCh38 | Deletion |
|
| esv2758993 | Chr.1:207384550 - 207751722 on Build GRCh38 | Gain+Loss |
|
| esv3545666 | Chr.1:207509399 - 207692876 on Build GRCh38 | Deletion |
|
| nsv8746 | Chr.1:207666908 - 207674530 on Build GRCh38 | Loss |
|
| esv2722117 | Chr.1:207634418 - 207750705 on Build GRCh38 | Deletion |
|

Genomic Map