Assay Details
Target Gene Details
Entrez Gene ID: | 576 |
Gene Name: | adhesion G protein-coupled receptor B2 |
Gene Aliases: |
BAI2 |
Location: |
Chr.1:31727105-31764063 on Build GRCh38 |
Assay Gene Location: | Within Intron 28 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950385 | Chr.1:31655500 - 31800799 on Build GRCh38 | Deletion |
|
| nsv1007786 | Chr.1:31725560 - 31830924 on Build GRCh38 | Gain |
|
| nsv545953 | Chr.1:31699894 - 31749577 on Build GRCh38 | Loss |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map