Assay Details
Target Gene Details
Entrez Gene ID: | 6390 |
Gene Name: | succinate dehydrogenase complex iron sulfur subunit B |
Gene Aliases: |
CWS2, IP, PGL4, SDH, SDH1, SDH2, SDHIP |
Location: |
Chr.1:17018722-17054170 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHB | NM_003000.2 | NP_002991.2 | ||
| AK312056.1 | ||||
| BC007840.2 | AAH07840.1 | |||
| BF796275.1 | ||||
| D10245.1 | BAA01089.1 | |||
| DQ403007.1 | ||||
| M32246.1 | AAA35708.1 | |||
| U17248.1 | AAA81167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758925 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss |
|
| nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss |
|
| nsv834269 | Chr.1:16855283 - 17038531 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map