Assay Details
Target Gene Details
Entrez Gene ID: | 54576 |
Gene Name: | UDP glucuronosyltransferase family 1 member A8 |
Gene Aliases: |
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H |
Location: |
Chr.2:233617645-233773299 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGT1A8 | NM_019076.4 | NP_061949.3 | ||
| AF030310.1 | AAB84259.1 | |||
| AF462267.1 | AAL75963.1 | |||
| AF462268.1 | AAL75964.1 | |||
| AK313488.1 | ||||
| AY435143.1 | AAR95644.1 | |||
| DQ364251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762307 | Chr.2:233595877 - 233653333 on Build GRCh38 | Gain |
|
| nsv584704 | Chr.2:233618034 - 233660491 on Build GRCh38 | Loss |
|
| nsv961572 | Chr.2:233617748 - 233650941 on Build GRCh38 | Duplication |
|
| esv2721664 | Chr.2:233620664 - 233679344 on Build GRCh38 | Deletion |
|
| esv3893573 | Chr.2:233595452 - 233639688 on Build GRCh38 | Gain |
|
| esv2721663 | Chr.2:233617724 - 233636648 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map