Assay Details
Target Gene Details
Entrez Gene ID: | 54842 |
Gene Name: | major facilitator superfamily domain containing 6 |
Gene Aliases: |
MMR2, hMMR2 |
Location: |
Chr.2:190408355-190502315 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD6 | NM_017694.3 | NP_060164.3 | ||
| XM_005246654.2 | XP_005246711.1 | |||
| XM_005246656.3 | XP_005246713.1 | |||
| XM_011511368.1 | XP_011509670.1 | |||
| XM_017004392.1 | XP_016859881.1 | |||
| AB472299.1 | ||||
| AK000167.1 | BAA90987.1 | |||
| AK098085.1 | ||||
| AK127176.1 | BAC86869.1 | |||
| AK225240.1 | ||||
| BC050537.2 | AAH50537.2 |
Target Gene Details
Entrez Gene ID: | 100131211 |
Gene Name: | nuclear envelope integral membrane protein 2 |
Gene Aliases: |
TMEM194B |
Location: |
Chr.2:190454088-190607126 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEMP2 | XM_011510458.2 | XP_011508760.1 | ||
| XM_017003100.1 | XP_016858589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33134 | Chr.2:190454065 - 190500984 on Build GRCh38 | Gain |
|
| esv3593756 | Chr.2:190455167 - 190455940 on Build GRCh38 | Loss |
|

Genomic Map